Learn More
Biomatik Corporation Recombinant Interleukin 4 (IL4), 100UG
Supplier:  Biomatik Corporation RPU53798
Encompass_Preferred
Recombinant Chicken Interleukin 4 (IL4) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Chicken (Gallus). Target Name: Interleukin 4 (IL4). Target Synonyms: BSF1; BCGF1; B Cell Stimulatory Factor 1; Lymphocyte stimulatory factor 1; B Cell Growth Factor; Binetrakin; Pitrakinra. Accession Number: Q5W4U1. Gene ID: 416330. Expression Region: Leu26~Lys136. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5886