Learn More
Biomatik Corporation Recombinant Tumor Necrosis Factor Beta (TNFb), 100UG
Catalog No. 501505887
Encompass_Preferred
Recombinant Rat Tumor Necrosis Factor Beta (TNFb) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Tumor Necrosis Factor Beta (TNFb). Target Synonyms: LT; LTA; TNF-B; TNFSF1; Tumor Necrosis Factor Ligand Superfamily Member 1; Lymphotoxin Alpha. Accession Number: Q06332. Gene ID: 25008. Expression Region: Ser35~Ser191. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5887 Supplier Biomatik Corporation Supplier No. RPU53803
Recombinant Rat Tumor Necrosis Factor Beta (TNFb) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Tumor Necrosis Factor Beta (TNFb). Target Synonyms: LT; LTA; TNF-B; TNFSF1; Tumor Necrosis Factor Ligand Superfamily Member 1; Lymphotoxin Alpha. Accession Number: Q06332. Gene ID: 25008. Expression Region: Ser35~Ser191. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.