Learn More
Biomatik Corporation Recombinant Neuropathy Target Esterase (NTE), 100UG
Catalog No. 501505888
Encompass_Preferred
Recombinant Rat Neuropathy Target Esterase (NTE) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Neuropathy Target Esterase (NTE). Target Synonyms: PNPLA6; Sws; Patatin-Like Phospholipase Domain Containing 6. Accession Number: M0R715. Gene ID: 360753. Expression Region: Leu965~Thr1137. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5888 Supplier Biomatik Corporation Supplier No. RPU53878
Recombinant Rat Neuropathy Target Esterase (NTE) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Neuropathy Target Esterase (NTE). Target Synonyms: PNPLA6; Sws; Patatin-Like Phospholipase Domain Containing 6. Accession Number: M0R715. Gene ID: 360753. Expression Region: Leu965~Thr1137. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.