Learn More
Biomatik Corporation Recombinant Angiopoietin Like Protein 8 (ANGPTL8), 100UG
Supplier:  Biomatik Corporation RPU54027
Encompass_Preferred
Recombinant Mouse Angiopoietin Like Protein 8 (ANGPTL8) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Angiopoietin Like Protein 8 (ANGPTL8). Target Synonyms: RIFL; TD26; C19orf80; Lipasin; Betatrophin; Hepatocellular Carcinoma-Associated Protein TD26. Accession Number: Q8R1L8. Gene ID: 624219. Expression Region: Val16~Ala198. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5894