Learn More
Biomatik Corporation Recombinant Interferon Epsilon (IFNe), 100UG
Catalog No. 501505895
Encompass_Preferred
Recombinant Human Interferon Epsilon (IFNe) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interferon Epsilon (IFNe). Target Synonyms: IFN-E1. Accession Number: Q86WN2. Gene ID: 338376. Expression Region: Gln30~Arg208. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5895 Supplier Biomatik Corporation Supplier No. RPU54091
Recombinant Human Interferon Epsilon (IFNe) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interferon Epsilon (IFNe). Target Synonyms: IFN-E1. Accession Number: Q86WN2. Gene ID: 338376. Expression Region: Gln30~Arg208. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.