Learn More
Biomatik Corporation Recombinant Myeloperoxidase (MPO), 100UG
Supplier:  Biomatik Corporation RPU54162
Encompass_Preferred
Recombinant Rat Myeloperoxidase (MPO) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Myeloperoxidase (MPO). Accession Number: D3ZGE2. Expression Region: Val134~Gly263. Tag Info: N-terminal His Tag. Theoretical MW: 19.0kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5898