Learn More
Biomatik Corporation Recombinant Complement Factor I (CFI), 100UG
Supplier:  Biomatik Corporation RPU54258
Encompass_Preferred
Recombinant Mouse Complement Factor I (CFI) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Complement Factor I (CFI). Target Synonyms: CF-I; C3b-INA; FI; IF; KAF; Factor I; Konglutinogen-Activating Factor; C3b-Inactivator; C3B/C4B inactivator. Accession Number: Q61129. Gene ID: 12630. Expression Region: Arg358~Val603. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5899