Learn More
Biomatik Corporation Recombinant Interleukin 35 (IL35), 100UG
Supplier:  Biomatik Corporation RPU54474
Encompass_Preferred
Recombinant Mouse Interleukin 35 (IL35) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Interleukin 35 (IL35). Accession Number: P43431. Gene ID: 16159. Expression Region: Arg23~Ala215. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5906