Learn More
Biomatik Corporation Recombinant Mucin 5 Subtype B (MUC5B), 100UG
Supplier:  Biomatik Corporation RPU54506
Encompass_Preferred
Recombinant Bovine Mucin 5 Subtype B (MUC5B) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: Mucin 5 Subtype B (MUC5B). Target Synonyms: MUC5-B; MUC9; MG1; Oligomeric Mucus/Gel-Forming; Mucin 5,Subtype B,Tracheobronchial; Cervical mucin; High molecular weight salivary mucin MG1; Sublingual gland mucin. Accession Number: F2FB42. Expression Region: Gln6489~Tyr6632. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5907