Learn More
Biomatik Corporation Recombinant Cluster of Differentiation 40 Ligand (CD40L), 100UG
Supplier:  Biomatik Corporation RPU54734
Encompass_Preferred
Recombinant Simian Cluster Of Differentiation 40 Ligand (CD40L) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Simian. Target Name: Cluster Of Differentiation 40 Ligand (CD40L). Target Synonyms: CD154; TRAP; HIGM1; IGM; IMD3; TBAM; T-BAM; TNFSF5; Gp39; TNF Superfamily Member 5; Hyper-IgM Syndrome; TNF-Related Activation Protein; T-Cell B-Cell Activating Molecule. Accession Number: P63304. Gene ID: 574160. Expression Region: Asn180~Leu261. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5915