Learn More
Biomatik Corporation Eukaryotic Tumor Necrosis Factor Alpha (TNFa), 100UG
Supplier:  Biomatik Corporation RPU54756
Encompass_Preferred
Eukaryotic Mouse Tumor Necrosis Factor Alpha (TNFa) is a purified Eukaryotic Protein. Purity: >90% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Tumor Necrosis Factor Alpha (TNFa). Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Accession Number: P06804. Gene ID: 21926. Expression Region: Gly57~Leu235. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5918