Learn More
Biomatik Corporation Eukaryotic Interleukin 1 Beta (IL1b), 100UG
Supplier:  Biomatik Corporation RPU54831
Encompass_Preferred
Eukaryotic Human Interleukin 1 Beta (IL1b) is a purified Eukaryotic Protein. Purity: >95% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 1 Beta (IL1b). Target Synonyms: IL1-B; IL1-Beta; IL1F2; IL-1β; Interleukin-1 Family Member 2; Catabolin. Accession Number: P01584. Gene ID: 3553. Expression Region: Ala117~Ser269. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5924