Learn More
Biomatik Corporation Active Stem Cell Factor (SCF), 100UG
Supplier:  Biomatik Corporation RPU54872
Encompass_Preferred
Pig Stem Cell Factor (SCF), Active Protein is a purified Eukaryotic Protein. Purity: >95% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Stem Cell Factor (SCF). Target Synonyms: KITLG; MGF; SF; KL1; Kitl; FPH2; Kit-Ligand; Steel Factor; Mast Cell Growth Factor; Steel factor; Familial Progressive Hyperpigmentation 2. Accession Number: Q29030. Gene ID: 397509. Expression Region: Thr25~Ala191. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5926