Learn More
Biomatik Corporation Active Granzyme M (GZMM), 100UG
Supplier:  Biomatik Corporation RPU54892
Encompass_Preferred
Human Granzyme M (GZMM), Active Protein is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Granzyme M (GZMM). Target Synonyms: GZM-M; LMET1; MET1; Hu-Met-1; Met-ase; Lymphocyte Met Ase 1; Met-1 serine protease; Natural killer cell granular protease. Accession Number: P51124. Gene ID: 3004. Expression Region: Ile26~Ala257. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5927