Learn More
Biomatik Corporation Active Cold Inducible RNA Binding Protein (CIRBP), 100UG
Supplier:  Biomatik Corporation RPU54980
Encompass_Preferred
Human Cold Inducible RNA Binding Protein (CIRBP), Active Protein is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cold Inducible RNA Binding Protein (CIRBP). Target Synonyms: CIRP; A18HNRNP; Glycine-Rich RNA Binding Protein. Accession Number: Q14011. Gene ID: 1153. Expression Region: Met1~Glu172. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5928