Learn More
Biomatik Corporation Recombinant Mouse Annexin A1 (Anxa1), 100UG
Supplier:  Biomatik Corporation RPC20219
Encompass_Preferred
Recombinant Mouse Annexin A1 (Anxa1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Anxa1. Target Synonyms: Annexin I; Annexin-1; Calpactin II; Calpactin-2; Chromobindin-9; Lipocortin I; Phospholipase A2 inhibitory protein; p35. Accession Number: P10107. Expression Region: 2~346aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 40.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4669