Learn More
Biomatik Corporation Recombinant Stem Cell Factor (SCF), 100UG
Supplier:  Biomatik Corporation RPU55180
Encompass_Preferred
Recombinant Human Stem Cell Factor (SCF) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Stem Cell Factor (SCF). Target Synonyms: KITLG; MGF; SF; KL1; Kitl; FPH2; Kit-Ligand; Steel Factor; Mast Cell Growth Factor; Steel factor; Familial Progressive Hyperpigmentation 2. Accession Number: P21583. Gene ID: 4254. Expression Region: Glu26~Ala217. Tag Info: N-terminal His Tag. Theoretical MW: 27/29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5934