Learn More
Biomatik Corporation Recombinant Lactoperoxidase (LPO), 100UG
Supplier:  Biomatik Corporation RPU55201
Encompass_Preferred
Recombinant Human Lactoperoxidase (LPO) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lactoperoxidase (LPO). Target Synonyms: SPO; Salivary peroxidase. Accession Number: P22079. Gene ID: 4025. Expression Region: Phe366~Phe572. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5936