Learn More
Biomatik Corporation Recombinant Interleukin 23 (IL23), 100UG
Supplier:  Biomatik Corporation RPU55202
Encompass_Preferred
Recombinant Mouse Interleukin 23 Subunit Alpha (IL23a) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Interleukin 23 Subunit Alpha (IL23a). Target Synonyms: P19; IL23P19; SGRF. Accession Number: Q9EQ14. Gene ID: 83430. Expression Region: Arg20~Pro189. Tag Info: N-terminal His Tag. Theoretical MW: 15/16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5937