Learn More
Biomatik Corporation Recombinant Brain Natriuretic Peptide (BNP), 100UG
Supplier:  Biomatik Corporation RPU55221
Encompass_Preferred
Recombinant Mouse Brain Natriuretic Peptide (BNP) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Brain Natriuretic Peptide (BNP). Target Synonyms: GC-B; B-Type Natriuretic Peptide; Ventricular Natriuretic Peptide; Gamma-brain natriuretic peptide; Brain natriuretic peptide 32. Accession Number: P40753. Gene ID: 18158. Expression Region: Tyr27~Leu121. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5938