Learn More
Biomatik Corporation Recombinant Human Annexin A5 (ANXA5), 100UG
Supplier:  Biomatik Corporation RPC20220
Encompass_Preferred
Recombinant Human Annexin A5 (ANXA5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ANXA5. Target Synonyms: Anchorin CII; Annexin V; Annexin-5; Calphobindin I; CBP-IEndonexin II; Lipocortin V; Placental anticoagulant protein 4; PP4Placental anticoagulant protein I; PAP-I; Thromboplastin inhibitor; Vascular anticoagulant-alpha; VAC-alpha. Accession Number: P08758. Expression Region: 2~320aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 37.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4670