Learn More
Biomatik Corporation Recombinant Apolipoprotein B100 (APOB100), 100UG
Supplier:  Biomatik Corporation RPU55236
Encompass_Preferred
Recombinant Human Apolipoprotein B100 (APOB100) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Apolipoprotein B100 (APOB100). Target Synonyms: Apo-B100. Accession Number: P04114. Gene ID: 338. Expression Region: His3365~Glu3548. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5939