Learn More
Biomatik Corporation Recombinant Synuclein Alpha (SNCa), 100UG
Supplier:  Biomatik Corporation RPU55272
Encompass_Preferred
Recombinant Human Synuclein Alpha (SNCa) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Synuclein Alpha (SNCa). Target Synonyms: SNC-A; aSYN; PD1; PARK1; PARK4; NACP; Non-A Beta Component Of Alzheimer's Disease Amyloid Precursor Protein; Non-A4 component of amyloid precursor. Accession Number: P37840. Gene ID: 6622. Expression Region: Met1~Ala140. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5941