Learn More
Biomatik Corporation Recombinant Netrin 1 (Ntn1), 100UG
Supplier:  Biomatik Corporation RPU55301
Encompass_Preferred
Recombinant Rat Netrin 1 (Ntn1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Netrin 1 (Ntn1). Target Synonyms: NTN1L; Epididymis tissue protein Li 131P. Accession Number: Q924Z9. Expression Region: Pro313~Pro565. Tag Info: N-terminal His and GST Tag. Theoretical MW: 58kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5943