Learn More
Biomatik Corporation Recombinant Matrix Metalloproteinase 25 (MMP25), 100UG
Catalog No. 501505944
Encompass_Preferred
Recombinant Human Matrix Metalloproteinase 25 (MMP25) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Matrix Metalloproteinase 25 (MMP25). Target Synonyms: MT6-MMP; MMP25; MMPL1; MT-MMP6; Matrix Metallopeptidase Like 1; Leukolysin; Membrane-type matrix metalloproteinase 6; Membrane-type-6 matrix metalloproteinase. Accession Number: Q9NPA2. Gene ID: 64386. Expression Region: Tyr108~Ile313. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5944 Supplier Biomatik Corporation Supplier No. RPU55310
Recombinant Human Matrix Metalloproteinase 25 (MMP25) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Matrix Metalloproteinase 25 (MMP25). Target Synonyms: MT6-MMP; MMP25; MMPL1; MT-MMP6; Matrix Metallopeptidase Like 1; Leukolysin; Membrane-type matrix metalloproteinase 6; Membrane-type-6 matrix metalloproteinase. Accession Number: Q9NPA2. Gene ID: 64386. Expression Region: Tyr108~Ile313. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.