Learn More
Biomatik Corporation Recombinant Calprotectin (CALPRO), 100UG
Catalog No. 501505946
Encompass_Preferred
Recombinant Mouse Calprotectin (CALPRO) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Calprotectin (CALPRO). Target Synonyms: MRP-8/MRP-14; S100A8/A9. Accession Number: P27005; P31725. Expression Region: Met1~Glu89andMet1~Lys113. Tag Info: N-terminal His Tag. Theoretical MW: 11/15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5946 Supplier Biomatik Corporation Supplier No. RPU55396
Recombinant Mouse Calprotectin (CALPRO) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Calprotectin (CALPRO). Target Synonyms: MRP-8/MRP-14; S100A8/A9. Accession Number: P27005; P31725. Expression Region: Met1~Glu89andMet1~Lys113. Tag Info: N-terminal His Tag. Theoretical MW: 11/15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.