Learn More
Biomatik Corporation Recombinant Human Atrial natriuretic peptide receptor 1 (NPR1), partial, 100UG
Supplier:  Biomatik Corporation RPC20239
Encompass_Preferred
Recombinant Human Atrial natriuretic peptide receptor 1 (NPR1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NPR1. Target Synonyms: Atrial natriuretic peptide receptor type A; ANP-A; ANPR-A; NPR-AGuanylate cyclase A; GC-A. Accession Number: P16066. Expression Region: 33~473aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 50.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4675