Learn More
Biomatik Corporation Recombinant Mouse Prostate stem cell antigen (Psca), 100UG
Catalog No. 501504676
Encompass_Preferred
Recombinant Mouse Prostate stem cell antigen (Psca) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Psca. Target Synonyms: Psca; Prostate stem cell antigen. Accession Number: P57096. Expression Region: 21~95aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 10.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4676 Supplier Biomatik Corporation Supplier No. RPC20241
Recombinant Mouse Prostate stem cell antigen (Psca) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Psca. Target Synonyms: Psca; Prostate stem cell antigen. Accession Number: P57096. Expression Region: 21~95aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 10.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.