Learn More
Biomatik Corporation Recombinant Juniperus ashei Pectate lyase 1, 100UG
Supplier:  Biomatik Corporation RPC20250
Encompass_Preferred
Recombinant Juniperus ashei Pectate lyase 1 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Juniperus ashei (Ozark white cedar). Target Name: Juniperus ashei Pectate lyase 1. Target Synonyms: Pectate lyase 1; EC 4.2.2.2; Major pollen allergen Jun a 1; allergen Jun a 1. Accession Number: P81294. Expression Region: 22~367aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 39.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4681