Learn More
Biomatik Corporation Recombinant Streptococcus pneumoniae serotype 4 Pneumolysin (ply), 100UG
Supplier:  Biomatik Corporation RPC20251
Encompass_Preferred
Recombinant Streptococcus pneumoniae serotype 4 Pneumolysin (ply) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Streptococcus pneumoniae serotype 4 (strain ATCC BAA-334 / TIGR4). Target Name: ply. Target Synonyms: Thiol-activated cytolysin. Accession Number: P0C2J9. Expression Region: 2~471aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 54.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4682