Learn More
Biomatik Corporation Recombinant Clostridium botulinum Botulinum neurotoxin type B (botB), partial, 100UG
Supplier:  Biomatik Corporation RPC20252
Encompass_Preferred
Recombinant Clostridium botulinum Botulinum neurotoxin type B (botB), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Clostridium botulinum. Target Name: botB. Target Synonyms: Bontoxilysin-B. Accession Number: P10844. Expression Region: 1~427aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 51.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4683