Learn More
Biomatik Corporation Recombinant Bovine coronavirus Hemagglutinin-esterase (HE), 100UG
Supplier:  Biomatik Corporation RPC20253
Encompass_Preferred
Recombinant Bovine coronavirus Hemagglutinin-esterase (HE) is a purified Recombinant Protein; Coronavirus Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Bovine coronavirus (strain Mebus) (BCoV) (BCV). Target Name: HE. Target Synonyms: E3 glycoprotein. Accession Number: P15776. Expression Region: 19~424aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 47.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4684