Learn More
Biomatik Corporation Recombinant Mouse Carboxylesterase 1C (Ces1c), partial, 100UG
Supplier:  Biomatik Corporation RPC20256
Encompass_Preferred
Recombinant Mouse Carboxylesterase 1C (Ces1c), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ces1c. Target Synonyms: Liver carboxylesterase NLung surfactant convertasePES-N. Accession Number: P23953. Expression Region: 19~550aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 60.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4685