Learn More
Biomatik Corporation Recombinant Porphyromonas gingivalis Gingipain R1 (rgpA), partial, 100UG
Supplier:  Biomatik Corporation RPC20257
Encompass_Preferred
Recombinant Porphyromonas gingivalis Gingipain R1 (rgpA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Porphyromonas gingivalis. Target Name: rgpA. Target Synonyms: Arg-gingipain; Gingipain 1RGP-1. Accession Number: P28784. Expression Region: 228~720aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 56kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4686