Learn More
Biomatik Corporation Recombinant Toxocara canis 26KDA secreted antigen (TES-26), 100UG
Supplier:  Biomatik Corporation RPC20259
Encompass_Preferred
Recombinant Toxocara canis 26KDA secreted antigen (TES-26) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Toxocara canis (Canine roundworm). Target Name: TES-26. Target Synonyms: Toxocara excretory-secretory antigen 26; TES-26. Accession Number: P54190. Expression Region: 22~262aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4687