Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Alpha-hemolysin (hly), 100UG
Supplier:  Biomatik Corporation RPC20267
Encompass_Preferred
Recombinant Staphylococcus aureus Alpha-hemolysin (hly) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Staphylococcus aureus (strain NCTC 8325). Target Name: hly. Target Synonyms: Alpha-toxin. Accession Number: Q2G1X0. Expression Region: 27~319aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 35.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4691