Learn More
Biomatik Corporation Recombinant Human Cytosolic 5'-nucleotidase 1A (NT5C1A), 100UG
Catalog No. 501504693
Encompass_Preferred
Recombinant Human Cytosolic 5'-nucleotidase 1A (NT5C1A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NT5C1A. Target Synonyms: Cytosolic 5'-nucleotidase IA; cN-I; cN-IA. Accession Number: Q9BXI3. Expression Region: 1~368aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 43kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4693 Supplier Biomatik Corporation Supplier No. RPC20270
Recombinant Human Cytosolic 5'-nucleotidase 1A (NT5C1A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NT5C1A. Target Synonyms: Cytosolic 5'-nucleotidase IA; cN-I; cN-IA. Accession Number: Q9BXI3. Expression Region: 1~368aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 43kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.