Learn More
Biomatik Corporation Recombinant Human Delta-like protein 3 (DLL3), partial, 100UG
Supplier:  Biomatik Corporation RPC20271
Encompass_Preferred
Recombinant Human Delta-like protein 3 (DLL3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DLL3. Target Synonyms: Drosophila Delta homolog 3; Delta3. Accession Number: Q9NYJ7. Expression Region: 27~492aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 50.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4694