Learn More
Biomatik Corporation Recombinant Klebsiella oxytoca Carbepenem-hydrolyzing beta-lactamase KPC (bla), 100UG
Supplier:  Biomatik Corporation RPC20275
Encompass_Preferred
Recombinant Klebsiella oxytoca Carbapenem-hydrolyzing beta-lactamase KPC (bla) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Klebsiella oxytoca. Target Name: bla. Target Synonyms: Carbapenem-hydrolyzing beta-lactamase KPC-2. Accession Number: Q848S6. Expression Region: 25~293aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 44.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4695