Learn More
Biomatik Corporation Recombinant Human S-arrestin (SAG), 100UG
Supplier:  Biomatik Corporation RPC20290
Encompass_Preferred
Recombinant Human S-arrestin (SAG) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SAG. Target Synonyms: 48 kDa proteinRetinal S-antigenS-AGRod photoreceptor arrestin. Accession Number: P10523. Expression Region: 1~405aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 47.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4700