Learn More
Biomatik Corporation Recombinant Epstein-Barr virus Envelope glycoprotein GP350 (BLLF1), partial, 100UG
Supplier:  Biomatik Corporation RPC20293
Encompass_Preferred
Recombinant Epstein-Barr virus Envelope glycoprotein GP350 (BLLF1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4). Target Name: BLLF1. Target Synonyms: Membrane antigenMA. Accession Number: P03200. Expression Region: 1~493aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 68.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4701