Learn More
Biomatik Corporation Recombinant Mouse Platelet-derived growth factor D (Pdgfd), 100UG
Supplier:  Biomatik Corporation RPC20299
Encompass_Preferred
Recombinant Mouse Platelet-derived growth factor D (Pdgfd) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Pdgfd. Target Synonyms: Spinal cord-derived growth factor BSCDGF-BCleaved into the following 2 chains:Platelet-derived growth factor D, latent formPDGFD latent formPlatelet-derived growth factor D, receptor-binding formPDGFD receptor-binding form. Accession Number: Q925I7. Expression Region: 24~370aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 56.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4705