Learn More
Biomatik Corporation Recombinant Blattella germanica Allergen Bla g 4, 100UG
Supplier:  Biomatik Corporation RPC20302
Encompass_Preferred
Recombinant Blattella germanica Allergen Bla g 4 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Blattella germanica (German cockroach) (Blatta germanica). Target Name: Blattella germanica Allergen Bla g 4. Target Synonyms: Allergen Bla g IVAllergen: Bla g 4. Accession Number: P54962. Expression Region: 13~182aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 35.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4708