Learn More
Biomatik Corporation Recombinant Agrobacterium sp. 3-phosphoshikimate 1-carboxyvinyltransferase (aroA), 100UG
Supplier:  Biomatik Corporation RPC20303
Encompass_Preferred
Recombinant Agrobacterium sp. 3-phosphoshikimate 1-carboxyvinyltransferase (aroA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Agrobacterium sp. (strain CP4). Target Name: aroA. Target Synonyms: 5-enolpyruvylshikimate-3-phosphate synthaseEPSP synthaseEPSPS. Accession Number: Q9R4E4. Expression Region: 1~455aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 63.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4709