Learn More
Biomatik Corporation Recombinant Human D-amino-acid oxidase (DAO), 100UG
Supplier:  Biomatik Corporation RPC20032
Encompass_Preferred
Recombinant Human D-amino-acid oxidase (DAO) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DAO. Target Synonyms: D-amino-acid oxidase; DAAO; DAMOX; DAO; DAO1; EC 1.4.3.3; MGC35381; OXDA; OXDA_HUMAN. Accession Number: P14920. Expression Region: 1~347aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 55.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4548