Learn More
Biomatik Corporation Recombinant Human Interferon-induced helicase C domain-containing protein 1 (IFIH1), partial, 100UG
Catalog No. 501504715
Encompass_Preferred
Recombinant Human Interferon-induced helicase C domain-containing protein 1 (IFIH1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IFIH1. Target Synonyms: Clinically amyopathic dermatomyositis autoantigen 140 kDa. Accession Number: Q9BYX4. Expression Region: 700~1025aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 41.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4715 Supplier Biomatik Corporation Supplier No. RPC20312
Recombinant Human Interferon-induced helicase C domain-containing protein 1 (IFIH1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IFIH1. Target Synonyms: Clinically amyopathic dermatomyositis autoantigen 140 kDa. Accession Number: Q9BYX4. Expression Region: 700~1025aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 41.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.