Learn More
Biomatik Corporation Recombinant Human Patatin-like phospholipase domain-containing protein 2 (PNPLA2), 100UG
Supplier:  Biomatik Corporation RPC20313
Encompass_Preferred
Recombinant Human Patatin-like phospholipase domain-containing protein 2 (PNPLA2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PNPLA2. Target Synonyms: Adipose triglyceride lipaseCalcium-independent phospholipase A2DesnutrinIPLA2-zetaPigment epithelium-derived factorTTS2.2Transport-secretion protein 2TTS2. Accession Number: Q96AD5. Expression Region: 1~504aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 71.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4716