Learn More
Biomatik Corporation Recombinant Marmota monax Interferon gamma (IFNG), 100UG
Supplier:  Biomatik Corporation RPC20316
Encompass_Preferred
Recombinant Marmota monax Interferon gamma (IFNG) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Marmota monax (Woodchuck). Target Name: IFNG. Target Synonyms: IFN-gamma. Accession Number: O35735. Expression Region: 24~166aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 18.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4717