Learn More
Biomatik Corporation Recombinant Human T-cell immunoreceptor with Ig and ITIM domains (TIGIT), partial, 100UG
Catalog No. 501504722
Encompass_Preferred
Recombinant Human T-cell immunoreceptor with Ig and ITIM domains (TIGIT), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TIGIT. Target Synonyms: V-set and immunoglobulin domain-containing protein 9V-set and transmembrane domain-containing protein 3. Accession Number: Q495A1. Expression Region: 22~141aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 29.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4722 Supplier Biomatik Corporation Supplier No. RPC20323
Recombinant Human T-cell immunoreceptor with Ig and ITIM domains (TIGIT), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TIGIT. Target Synonyms: V-set and immunoglobulin domain-containing protein 9V-set and transmembrane domain-containing protein 3. Accession Number: Q495A1. Expression Region: 22~141aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 29.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.