Learn More
Biomatik Corporation Recombinant Bacteroides fragilis Fragilysin (btfP), 100UG
Catalog No. 501504723
Encompass_Preferred
Recombinant Bacteroides fragilis Fragilysin (btfP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bacteroides fragilis. Target Name: btfP. Target Synonyms: Enterotoxin. Accession Number: P54355. Expression Region: 26~405aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 58.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4723 Supplier Biomatik Corporation Supplier No. RPC20324
Recombinant Bacteroides fragilis Fragilysin (btfP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bacteroides fragilis. Target Name: btfP. Target Synonyms: Enterotoxin. Accession Number: P54355. Expression Region: 26~405aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 58.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.